Learn More
Stoelting™ Hudson (Ewald) Forceps
SDP
Catalog No. 10001220 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
1x2
Serrated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-220 1x2
10-001-219 Serrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-220 Supplier Stoelting™ Supplier No. 5210438
Only null left
Add to Cart
Add to Cart
Stoelting™ Hudson (Ewald) Forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure
  • Delicate hemostats have thin jaws with fine serrations
Specifications
For Use With (Application) Neuroscience
Length (English) 4.75 in.
Model 1x2