Learn More
US Plastic Corporation Hudson Extrusions Tubing, Natural Polypropylene, 1/8 in. ID, 1/4 in. OD, 0.062 in. Wall
SDP
Supplier:  US Plastic Corporation 58020
Encompass
Natural polypropylene tubing is ideal for transferring hot liquids or gases or where higher temperatures and pressures are expected. It is also used in manufacturing operations requiring extreme resistance to acids and alkalis.
  • Flexible, odorless, tasteless & non-toxic
  • Use compression fittings with this tubing
  • Hardness (Rockwell R): 100
  • Maximum temperature: 180°F
  • Maximum working pressure at 70°F: 272 psi
  • FDA-compliant resin/materials
Catalog No. NC3978025