Learn More
Promocell GmbH PromoCell HUF-c 500,000 Human Uterine Fibroblasts, cryopreserved
Catalog No. 50306012
Encompass
Primary Human Uterine Fibroblasts (HUF) are isolated from the uterine wall. During pregnancy, the uterine wall undergoes deep but reversible structural and biochemical changes. The uterine wall is the target of hormones and growth factors that act simultaneously and lead to extensive modifications of the extracellular matrix. This and other morphological and biochemical effects can be studied in vitro using HUF. Available formats:* Cryopreserved: Cryogenic vial containing 500.000 viable cells.
Catalog No. 50-306-012 Supplier Promocell GmbH Supplier No. C12385
Primary Human Uterine Fibroblasts (HUF) are isolated from the uterine wall. During pregnancy, the uterine wall undergoes deep but reversible structural and biochemical changes. The uterine wall is the target of hormones and growth factors that act simultaneously and lead to extensive modifications of the extracellular matrix. This and other morphological and biochemical effects can be studied in vitro using HUF. Available formats:* Cryopreserved: Cryogenic vial containing 500.000 viable cells.