Learn More
Medchemexpress LLC HuGAL-F2 1mg | 1mg
SDP
Catalog No. 5000324277
Encompass_Preferred
HuGAL-F2 is a human-derived antibody expressed in CHO cells targeting FGF2 HuGAL-F2 contains a huIgG2 heavy chain and a hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for HuGAL-F2 can refer to Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24277 Supplier Medchemexpress LLC Supplier No. HYP9905971MG
HuGAL-F2 is a human-derived antibody expressed in CHO cells targeting FGF2 HuGAL-F2 contains a huIgG2 heavy chain and a hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for HuGAL-F2 can refer to Human IgG1 kappa Isotype Control (HY-P99001)