Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hughes 16-Gallon Gravity Fed Eyewash Heated Jacket 120 V plug
GREENER_CHOICE

Catalog No. 19141124 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
Hughes 16 gal. Gravity Fed Eye Wash Stations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-141-124 Hughes 16 gal. Gravity Fed Eye Wash Stations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-141-124 Supplier Justrite Manufacturing Co Supplier No. 16GFEWBLKTHT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maintain a tepid temperature within the gravity fed eye wash station. The jacket be used in a cold or warm climate, it's simple to put on or take off the eye wash station

  • Whether in a cold or hot climate, our heated jacket keeps the 16 gallon eyewash station a tepid temperature
  • Constructed of heavyweight PVC cloth and double stitched to provide extra durability
  • Easily installed to fit our eye wash station

Specifications

Color Transparent
For Use With (Equipment) Hughes 16 gal. Gravity Fed Eye Wash Stations
Includes 1 Complete Unit
Material PVC-coated Polyester
Product Type Gravity Fed Eye Wash Heated Jacket
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.