Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Hughes 16-Gallon Gravity Fed Eyewash Insulated Jacket
GREENER_CHOICE

Catalog No. 19141123 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
Hughes 16 gal. Gravity Fed Eye Wash Stations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-141-123 Hughes 16 gal. Gravity Fed Eye Wash Stations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-141-123 Supplier Justrite™ Supplier No. 16GFEWBLKT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maintains a tepid temperature within the gravity fed eye wash station. The jacket be used in a cold or warm climate, it's simple to put on or take off the eye wash station

  • Whether in a cold or hot climate, our insulated jacket keeps the 16 gallon eyewash station a tepid temperature
  • Constructed of heavyweight PVC cloth and double stitched to provide extra durability
  • Easily installed to fit our eye wash station

Specifications

Color Transparent
For Use With (Equipment) Hughes 16 gal. Gravity Fed Eye Wash Stations
Includes One Complete Unit
Material PVC-coated Polyester
Product Type Gravity Fed Eye Wash Jacket
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.