The Hulk® HK5 Series safety glasses from MCR Safety offer superior eye protection with a modern single-lens design, providing an excellent horizontal and vertical field of vision. These glasses feature soft, permanently bonded TPR temple arms and a durable polycarbonate nose bridge for comfort and reliability, meeting ANSI/ISEA Z87.1 and CAN/CSA Z94.3 standards.
- Gray lens with Duramass Hard Coat
- Modern single-lens design for excellent horizontal and vertical field of vision
- Soft, permanently bonded TPR temple arms for secure above-the-ear grip
- Integral polycarbonate nose bridge
- UV420 lens resin protects against HEV light
- Provides 100% protection against ultraviolet rays
- Meets ANSI/ISEA Z87.1 and CAN/CSA Z94.3 standards
- TAA Compliant