Learn More
MSC Quantum Storage Hulk Hopper Stacking Bin, Polypropylene, Red, 1 ea
Catalog No. 5000515586
Encompass
This hopper stacking bin is designed for efficient organization and heavy-duty storage applications. Constructed from durable polypropylene, the container features a stackable design with a wide stacking ledge and anti-slide lock for secure, multi-unit organization. It is engineered for industrial environments, offering resistance to extreme cold and maintaining a waterproof, corrosion-free finish.
  • Heavy-duty front, back, and side grips for easy handling
  • Autoclavable up to 250°F
  • Stackable design with a wide stacking ledge and anti-slide lock
  • Waterproof and corrosion-free construction
  • Resistant to extreme cold
  • Load capacity of 150 lb.
Catalog No. 50-005-15586 Supplier MSC Supplier No. 91826081
This hopper stacking bin is designed for efficient organization and heavy-duty storage applications. Constructed from durable polypropylene, the container features a stackable design with a wide stacking ledge and anti-slide lock for secure, multi-unit organization. It is engineered for industrial environments, offering resistance to extreme cold and maintaining a waterproof, corrosion-free finish.
  • Heavy-duty front, back, and side grips for easy handling
  • Autoclavable up to 250°F
  • Stackable design with a wide stacking ledge and anti-slide lock
  • Waterproof and corrosion-free construction
  • Resistant to extreme cold
  • Load capacity of 150 lb.