Learn More
MSC Quantum Storage Hopper Stacking Bin, Polypropylene, 29-7/8 in. x 18-1/4 in. x 12 in., Red, 1 ea.
Catalog No. 5000515122
Encompass
This hopper stacking bin is constructed from high-impact polypropylene for heavy-duty industrial and warehouse applications. Its design provides excellent strength, longevity, and resistance to wear, making it suitable for efficient storage, transport, and workflow optimization in various environments.
  • Manufactured from high-impact polypropylene
  • Stackable design featuring a wide stacking ledge and anti-slide lock to prevent forward shifting
  • Resistant to extreme cold and corrosion-free with waterproof properties
  • Autoclavable up to 250°F
  • Equipped with rear handle for easy grip
  • Inside dimensions are 27-1/2 in. long by 16 in. wide by 11-3/4 in. high
  • Compatible with optional clear plastic windows and dividers for organization
  • Made in the USA
Catalog No. 50-005-15122 Supplier MSC Supplier No. 91826057
This hopper stacking bin is constructed from high-impact polypropylene for heavy-duty industrial and warehouse applications. Its design provides excellent strength, longevity, and resistance to wear, making it suitable for efficient storage, transport, and workflow optimization in various environments.
  • Manufactured from high-impact polypropylene
  • Stackable design featuring a wide stacking ledge and anti-slide lock to prevent forward shifting
  • Resistant to extreme cold and corrosion-free with waterproof properties
  • Autoclavable up to 250°F
  • Equipped with rear handle for easy grip
  • Inside dimensions are 27-1/2 in. long by 16 in. wide by 11-3/4 in. high
  • Compatible with optional clear plastic windows and dividers for organization
  • Made in the USA