Learn More
MSC Quantum Storage Hulk Hopper Stacking Bin, Polypropylene, Red, 1 ea
Catalog No. 5000511069
Encompass
This hopper stacking bin provides a durable and efficient storage solution for organizing items in industrial or commercial environments. Constructed from high-quality polypropylene, this rectangular container is designed for secure stacking to maximize vertical storage space.
  • Manufactured from robust polypropylene material
  • Hopper stacking bin style for organized storage
  • Designed for secure stacking capability
  • Latex free construction
  • Includes compatible divider slot for partition usage
  • Overall dimensions are 16-1/2 in. W x 35-7/8 in. L x 17-1/2 in. H
  • Made in the USA
Catalog No. 50-005-11069 Supplier MSC Supplier No. 91826115
This hopper stacking bin provides a durable and efficient storage solution for organizing items in industrial or commercial environments. Constructed from high-quality polypropylene, this rectangular container is designed for secure stacking to maximize vertical storage space.
  • Manufactured from robust polypropylene material
  • Hopper stacking bin style for organized storage
  • Designed for secure stacking capability
  • Latex free construction
  • Includes compatible divider slot for partition usage
  • Overall dimensions are 16-1/2 in. W x 35-7/8 in. L x 17-1/2 in. H
  • Made in the USA