Learn More
Biotang Inc Human 6-keto-prostaglandin F1a,6-K-PGF1a ELISA Kit, 96T
Catalog No. 50152562 Shop All BIOTANG Inc Products
Encompass_Preferred
Human 6-keto-prostaglandin F1a,6-K-PGF1a ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.
Catalog No. 50-152-562 Supplier Biotang Inc Supplier No. HU9355
Human 6-keto-prostaglandin F1a,6-K-PGF1a ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.