Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems ACROBiosystems Human Activin A (INHBA) Protein, Premium Grade, Recombinant CHO-Expressed, Gly 311-Ser 426, 50 μg (ACA-H5314)

Catalog No. NC3716168
Encompass_Preferred

Recombinant human Activin A (INHBA) protein corresponding to the mature inhibin beta A region (Gly 311-Ser 426), produced in CHO cells and supplied lyophilized for use in cell differentiation, receptor-binding, and reporter assays.

  • Recombinant CHO-expressed mature Activin A (Gly 311-Ser 426).
  • Supplied lyophilized from PBS with trehalose as protectant.
  • Purity greater than 95% as determined by SDS-PAGE.
  • Migrates at approximately 13 kDa (±3 kDa) under reducing SDS-PAGE.
  • Specific activity >5.00 x 10^2 IU/mg, calibrated to WHO reference reagent.
  • Suitable for endoderm induction, receptor-binding assays, and cell-based reporter assays.
  • Store lyophilized at ≤ -20°C; after reconstitution store at ≤ -70°C and avoid repeated freeze-thaw cycles.
  • Available in research sizes including 50 μg.

Catalog No. NC3716168 Supplier ACROBiosystems Supplier No. ACAH531450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Activin A (INHBA) protein corresponding to the mature inhibin beta A region (Gly 311-Ser 426), produced in CHO cells and supplied lyophilized for use in cell differentiation, receptor-binding, and reporter assays.

  • Recombinant CHO-expressed mature Activin A (Gly 311-Ser 426).
  • Supplied lyophilized from PBS with trehalose as protectant.
  • Purity greater than 95% as determined by SDS-PAGE.
  • Migrates at approximately 13 kDa (±3 kDa) under reducing SDS-PAGE.
  • Specific activity >5.00 x 10^2 IU/mg, calibrated to WHO reference reagent.
  • Suitable for endoderm induction, receptor-binding assays, and cell-based reporter assays.
  • Store lyophilized at ≤ -20°C; after reconstitution store at ≤ -70°C and avoid repeated freeze-thaw cycles.
  • Available in research sizes including 50 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.