Learn More
Angio Proteomie Human Alveolar Epithelial Cells, Frozen Vial, > 5e5 cells/vial, Passage 2, Population Doubling Levels > 15, Research Use Only, -80C short-term storage, Liquid nitrogen long-term storage
SDP
Catalog No. NC3793086
Encompass
Human Alveolar Epithelial Cells, Frozen Vial, > 5e5 cells/vial, Passage 2, Population Doubling Levels > 15, Research Use Only, -80C short-term storage, Liquid nitrogen long-term storage
Catalog No. NC3793086 Supplier Angio Proteomie Supplier No. CAP0208
Human Alveolar Epithelial Cells, Frozen Vial, > 5e5 cells/vial, Passage 2, Population Doubling Levels > 15, Research Use Only, -80C short-term storage, Liquid nitrogen long-term storage