Learn More
ACROBiosystems Recombinant Protein;Human B7-1 / CD80 Protein;HEK293;1MG;B71,CD80,B7,B7-1,B7.1,BB1,CD28LG,CD28LG1,LAB7
Supplier:  ACROBiosystems B71H52281MG
Encompass_Preferred
Recombinant Protein;Human B7-1 / CD80 Protein;1MG;Human B7-1, His Tag (B71-H5228) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # P33681-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 43-60 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;Immobilized Human CTLA-4, Fc Tag (Cat. No. CT4-H5255) at 10 μg/mL (100 μL/well) can bind Human B7-1, His Tag (Cat. No. B71-H5228) with a linear range of 0.156-1.25 μg/mL (QC tested).
Catalog No. 50-101-7230