Learn More
ACROBiosystems Recombinant Protein;Human CD3 epsilon Protein, His Tag & Fc Tag;HEK293;100UG;CDE,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3 epsilon ,CDE-H822R
Catalog No. 501017503
Encompass_Preferred
Recombinant Protein;Human CD3 epsilon Protein, His Tag & Fc Tag;100UG;Human CD3E, His Tag, Fc Tag (CDE-H5256) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # NP_000724.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 39.2 kDa. The protein migrates as 44-54 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti Human CD3 MAb, mouse IgG1 (Clone # SP34-2) at 2 μg/mL (100 μL/well) can bind Human CD3E, His Tag, Fc Tag (Cat. No. CDE-H5256) with a linear range of 0.1-3 ng/mL (Routinely tested).
Catalog No. 50-101-7503 Supplier ACROBiosystems Supplier No. CDEH5256100UG
Recombinant Protein;Human CD3 epsilon Protein, His Tag & Fc Tag;100UG;Human CD3E, His Tag, Fc Tag (CDE-H5256) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # NP_000724.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 39.2 kDa. The protein migrates as 44-54 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti Human CD3 MAb, mouse IgG1 (Clone # SP34-2) at 2 μg/mL (100 μL/well) can bind Human CD3E, His Tag, Fc Tag (Cat. No. CDE-H5256) with a linear range of 0.1-3 ng/mL (Routinely tested).