Learn More
ACROBiosystems Recombinant Protein;Human CD3 epsilon & CD3 gamma Heterodimer Protein;HEK293;50UG;CDG,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3 epsilon,CD3G,CD3-GAMMA,IMD17,T3G,CD3 gamma,TCR gamma,CD3E & CD3G
Catalog No. 501138240
Encompass_Preferred
Recombinant Protein;Human CD3 epsilon & CD3 gamma Heterodimer Protein;50UG;Human CD3E & CD3G, His Tag, Fc Tag, Flag Tag (CDG-H52W9) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # NP_000724.1 (CD3E) & AAI13831 (CD3G)).;The heterodimer Human CD3 epsilon & CD3 gamma is produced by co-expression of CD3E and CD3G, has a calculated MW of 39.2 kDa (CD3E) and 38.6 kDa (CD3G). Subunit CD3E is fused with a human IgG1 Fc tag at the C-terminus, followed by a polyhistidine tag, and subunit CD3G is fused with a human IgG1 Fc tag at the C-terminus, followed by a Flag tag. The predicted N-terminus is Asp 23 (CD3E) & Gln 23 (CD3G). The reducing (R) heterodimer protein migrates as 40-50 kDa due to glycosylation.;Immobilized OKT3 MAb at 10μg/mL (100 μL/well) can bind Human CD3E & CD3G (Cat. No. CDG-H52W9) with a linear range of 0.3-5 μg/mL (Routinely tested).;Immobilized Mouse Anti Human CD3 at 5 μg/mL (100 μL/well) can bind Huma
Catalog No. 50-113-8240 Supplier ACROBiosystems Supplier No. CDGH52W950UG
Recombinant Protein;Human CD3 epsilon & CD3 gamma Heterodimer Protein;50UG;Human CD3E & CD3G, His Tag, Fc Tag, Flag Tag (CDG-H52W9) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # NP_000724.1 (CD3E) & AAI13831 (CD3G)).;The heterodimer Human CD3 epsilon & CD3 gamma is produced by co-expression of CD3E and CD3G, has a calculated MW of 39.2 kDa (CD3E) and 38.6 kDa (CD3G). Subunit CD3E is fused with a human IgG1 Fc tag at the C-terminus, followed by a polyhistidine tag, and subunit CD3G is fused with a human IgG1 Fc tag at the C-terminus, followed by a Flag tag. The predicted N-terminus is Asp 23 (CD3E) & Gln 23 (CD3G). The reducing (R) heterodimer protein migrates as 40-50 kDa due to glycosylation.;Immobilized OKT3 MAb at 10μg/mL (100 μL/well) can bind Human CD3E & CD3G (Cat. No. CDG-H52W9) with a linear range of 0.3-5 μg/mL (Routinely tested).;Immobilized Mouse Anti Human CD3 at 5 μg/mL (100 μL/well) can bind Huma