Learn More
Lonza Walkersville Human Cryopreserved Kupffer Cells, ≥0.5 million cells/vial, Passage 0, Viability ≥70%, Cryo vial, liquid nitrogen storage or -180 °C
Catalog No. NC2832707
Encompass
Human Cryopreserved Kupffer Cells, ≥0.5 million cells/vial, Passage 0, Viability ≥70%, Cryo vial, liquid nitrogen storage or -180 °C
Catalog No. NC2832707 Supplier Lonza Walkersville Supplier No. HLKC500K
Human Cryopreserved Kupffer Cells, ≥0.5 million cells/vial, Passage 0, Viability ≥70%, Cryo vial, liquid nitrogen storage or -180 °C