Learn More
Discovery Life Sciences HUM CYP2A6+REDUCTASE+B5 0.5NMO
SDP
Catalog No. NC9685147
Encompass
Human CYP2A6 + P450 Reductase + Cytochrome b5 SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP2A6 expressed from human CYP2A6 cDNA using a baculovirus expression system
Catalog No. NC9685147 Supplier Discovery Life Sciences Supplier No. 456254
Human CYP2A6 + P450 Reductase + Cytochrome b5 SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP2A6 expressed from human CYP2A6 cDNA using a baculovirus expression system