Learn More
Discovery Life Sciences HUM CYP2B6+REDUCTASE+B5 0.5NMO
SDP
Catalog No. NC9685148
Encompass
Human CYP2B6 + P450 Reductase + Cytochromebeta5 SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL 7-Ethoxy-4-trifluoromethylcoumarin deethylase activity has replaced 7-ethoxycoumarin deethylase activity for this enzyme
Catalog No. NC9685148 Supplier Discovery Life Sciences Supplier No. 456255
Human CYP2B6 + P450 Reductase + Cytochromebeta5 SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL 7-Ethoxy-4-trifluoromethylcoumarin deethylase activity has replaced 7-ethoxycoumarin deethylase activity for this enzyme