Learn More
Discovery Life Sciences HUM CYP2C19+REDUCTASE+B5 0.5NM
SDP
Catalog No. NC9496284
Encompass
Human CYP2C19 + P450 Reductase + Cytochrome b5 SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP2C19 expressed from human CYP2C19 cDNA using a baculovirus expression system
Catalog No. NC9496284 Supplier Discovery Life Sciences Supplier No. 456259
Human CYP2C19 + P450 Reductase + Cytochrome b5 SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP2C19 expressed from human CYP2C19 cDNA using a baculovirus expression system