Learn More
Discovery Life Sciences HUM CYP2C8+REDUCTASE+B5 0.5NMO
SDP
Catalog No. NC9496282
Encompass
Human CYP2C8 + P450 Reductase + Cytochrome b5 SUPERSOMES, 0.5 nmole cytochrome P450 in 0.5 ml. Store at -80degreeC Catalytic assay: paclitaxel-6(alpha)-hydroxylase activity, representative catalytic activity: 2,200 pmole/(mg x min)
Catalog No. NC9496282 Supplier Discovery Life Sciences Supplier No. 456252
Human CYP2C8 + P450 Reductase + Cytochrome b5 SUPERSOMES, 0.5 nmole cytochrome P450 in 0.5 ml. Store at -80degreeC Catalytic assay: paclitaxel-6(alpha)-hydroxylase activity, representative catalytic activity: 2,200 pmole/(mg x min)