Learn More
Discovery Life Sciences HUM CYP2C91(ARG144)+REDUCTASE
SDP
Catalog No. NC9496283
Encompass
Human CYP2C91 + P450 Reductase + Cytochrome b5 SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP2C91 expressed from human CYP2C91 cDNA using a baculovirus expression system
Catalog No. NC9496283 Supplier Discovery Life Sciences Supplier No. 456258
Human CYP2C91 + P450 Reductase + Cytochrome b5 SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP2C91 expressed from human CYP2C91 cDNA using a baculovirus expression system