Learn More
Discovery Life Sciences HUM CYP2E1+REDUCTASE+B5 1NMOLE
SDP
Catalog No. NC9550136
Encompass
Human CYP2E1 + P450 Reductase + Cytochrome b5 SUPERSOMES 1.0 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP2E1 which is expressed from human CYP2E1 cDNA using a baculovirus expression system
Catalog No. NC9550136 Supplier Discovery Life Sciences Supplier No. 456206
Human CYP2E1 + P450 Reductase + Cytochrome b5 SUPERSOMES 1.0 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP2E1 which is expressed from human CYP2E1 cDNA using a baculovirus expression system