Learn More
Raybiotech Inc Quantibody Human Cytokine Array 1000 Kit, multiplexed, semi-quantitative, glass slide antibody array, 50 Samples, 10 μL per array
Catalog No. NC3761685
Encompass_Preferred
Quantibody Human Cytokine Array 1000 Kit, multiplexed, semi-quantitative, glass slide antibody array, 50 Samples, 10 μL per array
Catalog No. NC3761685 Supplier Raybiotech Inc Supplier No. QAHCAA10004
Add to Cart
Add to Cart
Request a Quote
Quantibody Human Cytokine Array 1000 Kit, multiplexed, semi-quantitative, glass slide antibody array, 50 Samples, 10 μL per array