Learn More
Abcam Human Adiponectin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1087861X96TESTS
Encompass_Preferred
Human Adiponectin ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Adiponectin in Human in Saliva, Urine, Plasma, Cell culture supernatant, Milk, Serum, Cerebral Spinal Fluid samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2319808