Learn More
Sciencell Research Laboratories Human Esophageal Epithelial Cells, Cell Only
Supplier:  Sciencell Research Laboratories 2720
Encompass
HEsEpiC from ScienCell Research Laboratories are isolated from the human esophagus. HEsEpiC are cryopreserved at passage one and delivered frozen. Each vial contains >5 x 10^5 cells in 1 ml volume.
Catalog No. NC1705562