Learn More
ACROBiosystems Recombinant Protein;Human Fc epsilon RI alpha Protein;HEK293;100UG;FCA,FCER1A,FCE1A,FcERI
Supplier:  ACROBiosystems FCAH5228100UG
Encompass_Preferred
Recombinant Protein;Human Fc epsilon RI alpha Protein;100UG;Human FCER1A, His Tag (FCA-H5228) is expressed from human 293 cells (HEK293). It contains AA Val 26 - Gln 205 (Accession # P12319-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.9 kDa. The protein migrates as 37-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Immunoglobulin E, Human Plasma at 5 μg/mL (100 μL/well) can bind Human FCER1A, His Tag (Cat. No. FCA-H5228) with a linear range of 0.4-3 ng/mL (QC tested).
Catalog No. 50-101-7784