Learn More
ACROBiosystems Recombinant Protein;Human G-CSF R / CD114 Protein, Fc Tag;HEK293;200UG;GCR,CSF3R,CD114,GCSFR,GCSF-R,G-CSF-R,CSF-3R
Supplier:  ACROBiosystems GCRH5250200UG
Encompass_Preferred
Recombinant Protein;Human G-CSF R / CD114 Protein, Fc Tag;200UG;Human G-CSF R, Fc Tag (GCR-H5250) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession # NP_000751.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 92.8 kDa. The protein migrates as 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human G-CSF, Tag Free (Cat. No. GCF-H5214) at 5 μg/mL (100 μL/well) can bind Human G-CSF R, Fc Tag (Cat. No. GCR-H5250) with a linear range of 0.5-5 ng/mL (QC tested).
Catalog No. 50-101-7869