Learn More
Enzo Life Sciences Human growth hormone (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT1890010
Encompass_Preferred
Alternative name: Somatotropin, GH1, HGH. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized in PBS. Source: Produced in HEK cells. The human growth hormone (aa 27-217) is fused at the N-terminus to a FLAG-tag followed by an 8 AA linker. Long Term Storage: -20°C.
Catalog No. 50-201-3976