Learn More
Abcam Human H2A.X (phospho S139) In-Cell ELISA Kit (IR), 1 x 96 Tests
Supplier:  Abcam AB1313821X96TESTS
Encompass_Preferred
Human H2A.X (phospho S139) In-Cell ELISA Kit (IR) is Cell-based (qualitative) ELISA for the measurement of Human H2A.X (phospho S139) (IR) production in Human in Suspension cells, Adherent cells samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2256472