Learn More
Lonza Walkersville Human Hepatocytes, Cryopreserved, Plateable
Catalog No. NC2152986
Encompass
Cryopreserved Human Hepatocytes, Plateable format are tested for high viability and healthy culture duration of at least 5 days in 2D sandwich culture. The General Purpose plateable hepatocytes are ideal for disease modeling, genomics applications, and other applications where high quality plated hepatocytes are required.The General Purpose hepatocytes should not be used in drug-drug interaction studies. For drug-drug interaction studies, please refer to the Induction Qualified hepatocytes (HUCPI).Benefits:Precharacterized for appropriate hepatocyte morphologyLarge inventory of multiple donorsUse for 3D culture applications, look for Verified for Spheroids Hepatocyte batches Tested for long-term applications and 96-well compatabilityApplications:Disease modelingGenomicsXenotransplantationContent:1 x Cryopreserved ampule HUCPG containing >5 million viable cells
This item is not returnable. View return policy
Catalog No. NC2152986 Supplier Lonza Walkersville Supplier No. HUCPG
Cryopreserved Human Hepatocytes, Plateable format are tested for high viability and healthy culture duration of at least 5 days in 2D sandwich culture. The General Purpose plateable hepatocytes are ideal for disease modeling, genomics applications, and other applications where high quality plated hepatocytes are required.The General Purpose hepatocytes should not be used in drug-drug interaction studies. For drug-drug interaction studies, please refer to the Induction Qualified hepatocytes (HUCPI).Benefits:Precharacterized for appropriate hepatocyte morphologyLarge inventory of multiple donorsUse for 3D culture applications, look for Verified for Spheroids Hepatocyte batches Tested for long-term applications and 96-well compatabilityApplications:Disease modelingGenomicsXenotransplantationContent:1 x Cryopreserved ampule HUCPG containing >5 million viable cells