Learn More
Biotang Inc Human Hexokinase,HK ELISA Kit, 96T
Supplier:  Biotang Inc HU8926
Encompass_Preferred
Human Hexokinase,HK ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.
Catalog No. 50-153-241