Learn More
ACROBiosystems Recombinant Protein;Human HSA Protein;HEK293;500UG;HSA,HSA,ALB,Serum Albumin
Supplier: ACROBiosystems HSAH5220500UG
Recombinant Protein;Human HSA Protein;500UG;Human HSA, His Tag (HSA-H5220) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Leu 609 (Accession # NP_000468).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.3 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.