Learn More
ACROBiosystems Recombinant Protein;ActiveMax Recombinant Human IFN-gamma /IFNG;HEK293;100UG;IFG,IFN-,IFNG,IFN,IFG,IFI,Interferon-gamma ,Interferon-,Interferon,Interferon gamma,IFN gamma,IFNgamma
Catalog No. 501017999
Encompass_Preferred
Recombinant Protein;ActiveMax Recombinant Human IFN-gamma /IFNG;100UG;ActiveMax Human IFN-gamma, Tag Free (IFG-H4211) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Gln 166 (Accession # AAH70256).;This protein carries no "tag". The protein has a calculated MW of 16.8 kDa. The protein migrates as 17 kDa,20 kDa and 24 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;The bio-activity of ActiveMax Human IFN-gamma, Tag Free (Cat. No. IFG-H4211) was determined by dose-dependent inhibition of the proliferation of HT-29 cells. The EC50 for this effect is typically 0.41-0.74 ng/mL (Routinely tested).
Catalog No. 50-101-7999 Supplier ACROBiosystems Supplier No. IFGH4211100UG
Recombinant Protein;ActiveMax Recombinant Human IFN-gamma /IFNG;100UG;ActiveMax Human IFN-gamma, Tag Free (IFG-H4211) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Gln 166 (Accession # AAH70256).;This protein carries no "tag". The protein has a calculated MW of 16.8 kDa. The protein migrates as 17 kDa,20 kDa and 24 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.;The bio-activity of ActiveMax Human IFN-gamma, Tag Free (Cat. No. IFG-H4211) was determined by dose-dependent inhibition of the proliferation of HT-29 cells. The EC50 for this effect is typically 0.41-0.74 ng/mL (Routinely tested).