Learn More
ACROBiosystems Recombinant Protein;Human IL-1RL1 / ST2 Protein;HEK293;1MG;IL1,IL1RL1,DER4,FIT-1,IL33R,ST2,ST2L,ST2V,T1,IL-1RL1,ST-2,IL-33R,ST-2L
Supplier:  ACROBiosystems IL1H52291MG
Encompass_Preferred
Recombinant Protein;Human IL-1RL1 / ST2 Protein;1MG;Human IL-1RL1, His Tag (IL1-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Phe 328 (Accession # Q01638-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL33 at 5 μg/mL (100 μL/well) can bind Human IL-1RL1, His Tag (Cat. No. IL1-H5229) with a linear range of 63-500 ng/mL (Routinely tested).
Catalog No. 50-101-8040