Learn More
ACROBiosystems Recombinant Protein;Human Integrin alpha V beta 8 (ITGAV&ITGB8) Heterodimer Protein;HEK293;100UG;IT8,Integrin alpha V beta 8,ITGAVB8,ITGAV&ITGB8
Supplier:  ACROBiosystems IT8H52W4100UG
Encompass_Preferred
Recombinant Protein;Human Integrin alpha V beta 8 (ITGAV&ITGB8) Heterodimer Protein;100UG;Human ITGAV & ITGB8 Heterodimer Protein (IT8-H52W4) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Glu 43 - Arg 684 (ITGB8) (Accession # NP_002201 (ITGAV) & NP_002205 (ITGB8)).;Human ITGAV & ITGB8 Heterodimer Protein, produced by co-expression of ITGAV and ITGB8, has a calculated MW of 113.0 kDa (ITGAV) and 76.5 kDa (ITGB8). Subunit ITGAV is fused with polyhistidine tag at the C-terminus and followed by a acidic tail and subunit ITGB8 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 145 kDa (ITGAV) and 100-116 kDa (ITGB8) respectively due to glycosylation.;Immobilized Human ITGAV & ITGB8 Heterodimer Protein (Cat. No. IT8-H52W4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human TGF-Beta 1, His Tag, Avi Tag with a linear range of 0.8-13 ng/mL (QC tested).;Immobilized Human Fibronectin at 10 μg/mL (100 μL/well) can bind
Catalog No. 50-113-8414