Learn More
ACROBiosystems Recombinant Protein;Human Jagged 1 / JAG1 Protein;HEK293;50UG;JA1,CD339,JAG1,Jagged1,hJ1,Jagged-1,JAGL1,JAG-1
Supplier:  ACROBiosystems JA1H52H950UG
Encompass_Preferred
Recombinant Protein;Human Jagged 1 / JAG1 Protein;50UG;Human Jagged 1, His Tag (JA1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gln 34 - Ser 1046 (Accession # P78504-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 112.3 kDa. The protein migrates as 130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-101-8168