Learn More
Diazyme Laboratories Human Kappa Free Light Chain Assay, Latex Enhanced Immunoturbidimetric, Serum Sample, Reagent 1: 22 mL, Reagent 2: 7 mL, 5 Calibrators (1.5 mL each), 10 min test time, LOD: 1.8 mg/L, LOQ: 2.9 mg/L, Storage: refrigeration
Catalog No. NC2222838
Encompass_Preferred
Human Kappa Free Light Chain Assay, Latex Enhanced Immunoturbidimetric, Serum Sample, Reagent 1: 22 mL, Reagent 2: 7 mL, 5 Calibrators (1.5 mL each), 10 min test time, LOD: 1.8 mg/L, LOQ: 2.9 mg/L, Storage: refrigeration
This item is not returnable. View return policy
Catalog No. NC2222838 Supplier Diazyme Laboratories Supplier No. DZ169AK
Human Kappa Free Light Chain Assay, Latex Enhanced Immunoturbidimetric, Serum Sample, Reagent 1: 22 mL, Reagent 2: 7 mL, 5 Calibrators (1.5 mL each), 10 min test time, LOD: 1.8 mg/L, LOQ: 2.9 mg/L, Storage: refrigeration