Learn More
Bio Rad Laboratories Bio-Plex Pro™ RBM Human Kidney Toxicity Panel 1 #171ATR1CK
Supplier:  Bio Rad Laboratories 171ATR1CK
Encompass
1 x 96-well, includes magnetic capture beads, detection antibodies, standards, controls, diluent, buffers, streptavidin-PE, flat bottom plate, and plate seals, for the detection of 6 kidney toxicity markers in human urine
Catalog No. NC3713086