Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories Human Lambda Free Light Chain Assay, Latex Enhanced Immunoturbidimetric method, Serum sample, Liquid Stable format, 20 μL sample volume, R1 Reagent 22 mL, R2 Reagent 7 mL, Calibrator 5 x 1.5 mL vials, Analytical range 3.5 - 4000 mg/L

Catalog No. NC2222840
Encompass_Preferred

The Diazyme Human Lambda Free Light Chain Assay is an advanced immunoturbidimetric analysis tool designed for the precise measurement of Lambda Free Light Chain (FLC) in human serum, essential for the diagnosis of plasma cell disorders like multiple myeloma. This assay not only promises optimal performance but is also ready-to-use, reducing preparation time and enhancing workflow efficiency in clinical settings.

  • Method: Latex enhanced immunoturbidimetric.
  • Sample type: Serum.
  • Sample volume: 20 μL.
  • Reagent format: Liquid stable.
  • Analytical measuring interval (Standard mode): 3.5 - 200 mg/L.
  • Analytical measuring interval (Extended mode): 3.5 - 4000 mg/L.
  • Linearity: 6.1 - 4000 mg/L.
  • Limit of detection (LOD): 2.8 mg/L.
  • Total precision: 1.1% - 11.4%.
  • Stored temperature: 2-8°C.

This item is not returnable. View return policy
Catalog No. NC2222840 Supplier Diazyme Laboratories Supplier No. DZ170AKY1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Diazyme Human Lambda Free Light Chain Assay is an advanced immunoturbidimetric analysis tool designed for the precise measurement of Lambda Free Light Chain (FLC) in human serum, essential for the diagnosis of plasma cell disorders like multiple myeloma. This assay not only promises optimal performance but is also ready-to-use, reducing preparation time and enhancing workflow efficiency in clinical settings.

  • Method: Latex enhanced immunoturbidimetric.
  • Sample type: Serum.
  • Sample volume: 20 μL.
  • Reagent format: Liquid stable.
  • Analytical measuring interval (Standard mode): 3.5 - 200 mg/L.
  • Analytical measuring interval (Extended mode): 3.5 - 4000 mg/L.
  • Linearity: 6.1 - 4000 mg/L.
  • Limit of detection (LOD): 2.8 mg/L.
  • Total precision: 1.1% - 11.4%.
  • Stored temperature: 2-8°C.

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.