Learn More
Biolegend LEGENDplex™ HU Macrophage/Microglia Panel (13-plex) w/ VbP
Catalog No. NC1875873
Encompass_Preferred
LEGENDplex™ HU Macrophage/Microglia Panel (13-plex) w/ VbP Apps: Multiplex - Panel; Size: 100 tests
This item is not returnable. View return policy
Catalog No. NC1875873 Supplier Biolegend Supplier No. 740503
Add to Cart
Add to Cart
LEGENDplex™ HU Macrophage/Microglia Panel (13-plex) w/ VbP Apps: Multiplex - Panel; Size: 100 tests