Learn More
Sciencell Research Laboratories Human Mammary Fibroblasts Cells only
Catalog No. NC2746105
Encompass
HMF from ScienCell Research Laboratories are isolated from human breast. HMF are cryopreserved at passage one and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume.
Catalog No. NC2746105 Supplier Sciencell Research Laboratories Supplier No. 7630
Add to Cart
Add to Cart
HMF from ScienCell Research Laboratories are isolated from human breast. HMF are cryopreserved at passage one and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume.