Learn More
Sciencell Research Laboratories Human Mammary Vascular Endothelial Cells
Catalog No. NC2746101
Encompass
HMVEC from ScienCell Research Laboratories are isolated from human breast. HMVEC are cryopreserved after purification and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume.
Catalog No. NC2746101 Supplier Sciencell Research Laboratories Supplier No. 7600
HMVEC from ScienCell Research Laboratories are isolated from human breast. HMVEC are cryopreserved after purification and delivered frozen. Each vial contains >5 x 105 cells in 1 ml volume.