Learn More
Kactus Mature TGF beta 3 Protein (TG3-HM00M) 100ug
Catalog No. NC3788017
Encompass
Recombinant Human Mature TGF beta 3 Protein is expressed from HEK293 without tag.It contains Ala301-Ser412.
Catalog No. NC3788017 Supplier Kactus Supplier No. TG3HM00M
Recombinant Human Mature TGF beta 3 Protein is expressed from HEK293 without tag.It contains Ala301-Ser412.