Learn More
ACROBiosystems Recombinant Protein;Human Mesothelin / MSLN (296-580) Protein, Fc Tag;HEK293;100UG;MSN,MSLN,Mesothelin,MPF
Supplier:  ACROBiosystems MSNH526X100UG
Encompass_Preferred
Recombinant Protein;Human Mesothelin / MSLN (296-580) Protein, Fc Tag;100UG;Human Mesothelin (296-580), Fc Tag (MSN-H526x) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 59.6 kDa. As a result of glycosylation, the protein migrates as 60-68 kDa under reducing (R) condition, and 120-130 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human Mesothelin (296-580), Fc Tag (Cat. No. MSN-H526x) at 0.5 μg/mL (100 μL/well) can bind Anti-Human MSLN Mab with a linear range of 0.8-3 ng/mL (QC tested).
Catalog No. 50-101-8302