Learn More
Meso Scale Discovery HUM MMP1 UltraSensitive Kit
SDP
Catalog No. 5000079652
Encompass
The Meso Scale Discovery MMP-1 Kit is a multiplex immunoassay that quantitatively measures MMP-1 in human serum plasma saliva and cell culture supernatant samples The kit uses electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrator solutions and assay diluent The kit can accurately quantify MMP-1 levels from 0 3 pg/mL to 10 000 pg/mL
Catalog No. 50-000-79652 Supplier Meso Scale Discovery Supplier No. K151FXC4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery MMP-1 Kit is a multiplex immunoassay that quantitatively measures MMP-1 in human serum plasma saliva and cell culture supernatant samples The kit uses electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrator solutions and assay diluent The kit can accurately quantify MMP-1 levels from 0 3 pg/mL to 10 000 pg/mL