Learn More
Meso Scale Discovery HUM MMP1 UltraSensitive Kit
SDP
Catalog No. 5000082037
Encompass
The Human MMP-1 Ultra-Sensitive Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) for quantifying matrix metalloproteinase 1 (MMP-1) in human serum plasma and cell culture supernatants It has a detection range of 0 3-1500 pg/mL offering high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and assay diluent providing an accurate tool for studying MMP-1 levels in research
Catalog No. 50-000-82037 Supplier Meso Scale Discovery Supplier No. K151FXC1
The Human MMP-1 Ultra-Sensitive Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) for quantifying matrix metalloproteinase 1 (MMP-1) in human serum plasma and cell culture supernatants It has a detection range of 0 3-1500 pg/mL offering high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and assay diluent providing an accurate tool for studying MMP-1 levels in research