Learn More
Meso Scale Discovery HUM MMP3 UltraSensitive Kit
SDP
Supplier:  Meso Scale Discovery K151FZC2
Encompass
The Meso Scale Discovery MMP-3 Kit is an electrochemiluminescent immunoassay for measuring matrix metalloproteinase 3 (MMP-3) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent Sensitivity is 0 3 ng/mL with a quantification limit of 10 000 pg/mL It requires a 2-hour incubation and detects MMP-3 in small samples aiding in extracellular matrix degradation studies
Catalog No. 50-000-78773