Learn More
Meso Scale Discovery HUM MMP3 UltraSensitive Kit
SDP
Catalog No. 5000081610
Encompass
The Meso Scale Discovery MMP-3 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of matrix metalloproteinase 3 (MMP-3) in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrator and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81610 Supplier Meso Scale Discovery Supplier No. K151FZC1
The Meso Scale Discovery MMP-3 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of matrix metalloproteinase 3 (MMP-3) in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrator and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL