Learn More
Invivogen HUM/ MURINE IL-1B SENSOR CELLS
SDP
Supplier:  Invivogen HKBIL1R
Encompass
Human & Murine IL-1 Sensor Cells 3-7 x 10e6 cells,HEK-Blue IL-1R cells were designed to detect bioactive human and murine cytokines (IL-1 and IL-1) by monitoring the activation of the NF-B and AP-1 pathways. Additionally, these cells detect bioactive IL-1 from cynomolgus monkeys, dogs, and rats. HEK-Blue IL-1R cells can detect IL-1 and IL-1 because these cytokines bind to the same receptor, IL-1R1.
Catalog No. NC1606826